Anti-NEUROD1 Mouse Monoclonal Antibody [clone: 6E9G12]

Supplier: GeneTex
GTX60691
10169-280EA 442.38 USD
10169-280
Anti-NEUROD1 Mouse Monoclonal Antibody [clone: 6E9G12]
Mouse Monoclonal Antibody [6E9G12] to NeuroD

Type: Primary
Antigen: NEUROD1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR