Anti-SLC27A5 Mouse Monoclonal Antibody [clone: 4B11C10]

Supplier: GeneTex
GTX60688
10169-274EA 582.91 USD
10169-274
Anti-SLC27A5 Mouse Monoclonal Antibody [clone: 4B11C10]
Antibodies
Mouse Monoclonal Antibody [4B11C10] to SLC27A5

Type: Primary
Antigen: SLC27A5
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR