Anti-CDH5 Mouse Monoclonal Antibody [clone: 3D5C7]

Supplier: GeneTex
GTX60681
10169-260EA 442 USD
10169-260
Anti-CDH5 Mouse Monoclonal Antibody [clone: 3D5C7]
Mouse Monoclonal Antibody [3D5C7] to CDH5

Type: Primary
Antigen: CDH5
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR