Anti-TGFBR3 Mouse Monoclonal Antibody [clone: 1C5H11]

Supplier: GeneTex
GTX60678
10169-254EA 551.93 USD
10169-254
Anti-TGFBR3 Mouse Monoclonal Antibody [clone: 1C5H11]
Antibodies
Mouse Monoclonal Antibody [1C5H11] to TGFBR3

Type: Primary
Antigen: TGFBR3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR