Anti-NES Mouse Monoclonal Antibody [clone: 1A9G12]

Supplier: GeneTex
GTX60677
10169-252EA 582.91 USD
10169-252
Anti-NES Mouse Monoclonal Antibody [clone: 1A9G12]
Antibodies
Mouse monoclonal antibody [1A9G12] to Nestin

Type: Primary
Antigen: NES
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR