Anti-SLC2A4 Mouse Monoclonal Antibody [clone: 3G10A3]

Supplier: GeneTex
GTX60674
10169-246EA 613.46 USD
10169-246
Anti-SLC2A4 Mouse Monoclonal Antibody [clone: 3G10A3]
Antibodies
Mouse Monoclonal Antibody [3G10A3] to SLC2A4

Type: Primary
Antigen: SLC2A4
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR