Anti-CDKN2A Mouse Monoclonal Antibody [clone: 1D7D2]

Supplier: GeneTex
GTX60663
10169-224EA 613.46 USD
10169-224
Anti-CDKN2A Mouse Monoclonal Antibody [clone: 1D7D2]
Antibodies
Mouse monoclonal antibody [1D7D2] to p14ARF

Type: Primary
Antigen: CDKN2A
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR