Anti-PPY Mouse Monoclonal Antibody [clone: 2G5D12]

Supplier: GeneTex
GTX60662
10169-222EA 582.91 USD
10169-222
Anti-PPY Mouse Monoclonal Antibody [clone: 2G5D12]
Antibodies
Mouse Monoclonal Antibody [2G5D12] to PPY

Type: Primary
Antigen: PPY
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR