Anti-ABCB5 Mouse Monoclonal Antibody [clone: 5H3C6]

Supplier: GeneTex
GTX60661
10169-220EA 582.91 USD
10169-220
Anti-ABCB5 Mouse Monoclonal Antibody [clone: 5H3C6]
Antibodies
Mouse Monoclonal Antibody [5H3C6] to ABCB5

Type: Primary
Antigen: ABCB5
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR