Anti-TRIP6 Mouse Monoclonal Antibody [clone: 6H4]

Supplier: GeneTex
GTX60653
10169-204EA 582.91 USD
10169-204
Anti-TRIP6 Mouse Monoclonal Antibody [clone: 6H4]
Antibodies
Mouse Monoclonal Antibody [6H4] to TRIP6

Type: Primary
Antigen: TRIP6
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR