Anti-FOXP1 Mouse Monoclonal Antibody [clone: 6E4]

Supplier: GeneTex
GTX60643
10169-184EA 582.91 USD
10169-184
Anti-FOXP1 Mouse Monoclonal Antibody [clone: 6E4]
Antibodies
Mouse Monoclonal Antibody [6E4] to FOXP1

Type: Primary
Antigen: FOXP1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR