Anti-POU3F2 Mouse Monoclonal Antibody [clone: 6H12]

Supplier: GeneTex
GTX60632
10169-162EA 582.91 USD
10169-162
Anti-POU3F2 Mouse Monoclonal Antibody [clone: 6H12]
Antibodies
Mouse Monoclonal Antibody [6H12] to POU3F2

Type: Primary
Antigen: POU3F2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR