Anti-CD5 Mouse Monoclonal Antibody [clone: 6A11]

Supplier: GeneTex
GTX60631
10169-160EA 582.91 USD
10169-160
Anti-CD5 Mouse Monoclonal Antibody [clone: 6A11]
Antibodies
Mouse Monoclonal Antibody [6A11] to CD5

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR