Anti-INCENP Mouse Monoclonal Antibody [clone: 3D2]

Supplier: GeneTex
GTX60619
10169-136EA 582.91 USD
10169-136
Anti-INCENP Mouse Monoclonal Antibody [clone: 3D2]
Antibodies
Mouse Monoclonal Antibody [3D2] to INCENP

Type: Primary
Antigen: INCENP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR