Anti-MYF6 Mouse Monoclonal Antibody [clone: 5D7]

Supplier: GeneTex
GTX60609
10169-116EA 582.91 USD
10169-116
Anti-MYF6 Mouse Monoclonal Antibody [clone: 5D7]
Antibodies
Mouse Monoclonal Antibody [5D7] to MYF6

Type: Primary
Antigen: MYF6
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR