Anti-PSMB8 Mouse Monoclonal Antibody [clone: 1A5]

Supplier: GeneTex
GTX60486
10168-498EA 582.91 USD
10168-498
Anti-PSMB8 Mouse Monoclonal Antibody [clone: 1A5]
Antibodies
Mouse Monoclonal Antibody [1A5] to PSMB8

Type: Primary
Antigen: PSMB8
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR