Anti-MMP1 Mouse Monoclonal Antibody [clone: 6A5]

Supplier: GeneTex
GTX60467
10168-348EA 582.91 USD
10168-348
Anti-MMP1 Mouse Monoclonal Antibody [clone: 6A5]
Antibodies
Mouse Monoclonal Antibody [6A5] to MMP1

Type: Primary
Antigen: MMP1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR