Anti-EPHA10 Mouse Monoclonal Antibody [clone: 2E8]

Supplier: GeneTex
GTX60439
10168-238EA 582.91 USD
10168-238
Anti-EPHA10 Mouse Monoclonal Antibody [clone: 2E8]
Antibodies
Mouse Monoclonal antibody [2E8] to EphA10

Type: Primary
Antigen: EPHA10
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR