Anti-MTHFR Mouse Monoclonal Antibody [clone: 5D3]

Supplier: GeneTex
GTX60435
10168-230EA 613.46 USD
10168-230
Anti-MTHFR Mouse Monoclonal Antibody [clone: 5D3]
Antibodies
Mouse Monoclonal Antibody [5D3] to MTHFR

Type: Primary
Antigen: MTHFR
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR