Anti-CEACAM5 Mouse Monoclonal Antibody [clone: 3G12]

Supplier: GeneTex
GTX60388
10168-154EA 582.91 USD
10168-154
Anti-CEACAM5 Mouse Monoclonal Antibody [clone: 3G12]
Antibodies
Mouse Monoclonal Antibody [3G12] to CEA

Type: Primary
Antigen: CEACAM5
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR