Anti-H2B Antibody Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX60346
10162-646EA 589.99 USD
10162-646
Anti-H2B Antibody Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Histone H2B

Type: Primary
Antigen: H2B antibody
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR