Anti-ESR1 Mouse Monoclonal Antibody [clone: GT9004]

Supplier: GeneTex
GTX60340
10162-634EA 457.31 USD
10162-634
Anti-ESR1 Mouse Monoclonal Antibody [clone: GT9004]
Antibodies
Mouse monoclonal antibody [GT9004] to Estrogen Receptor alpha

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR