Anti-Pigf Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX60318
10165-134EA 613.1 USD
10165-134
Anti-Pigf Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to PIGF

Type: Primary
Antigen: Pigf
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR