Anti-Fastkd5 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX60304
10165-120EA 613.1 USD
10165-120
Anti-Fastkd5 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to FASTKD5

Type: Primary
Antigen: Fastkd5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR