Anti-KDM5B Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX60284
10162-248EA 612.92 USD
10162-248
Anti-KDM5B Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to KDM5B

Type: Primary
Antigen: KDM5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR