Anti-ALK Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX59665
10165-168EA 550.56 USD
10165-168
Anti-ALK Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ALK

Type: Primary
Antigen: ALK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR