Anti-SLC3A2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX54716
10793-022EA 610.88 USD
10793-022
Anti-SLC3A2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to CD98

Type: Primary
Antigen: SLC3A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR