Anti-SEMG1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX54712
10793-014EA 610.69 USD
10793-014
Anti-SEMG1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Semenogelin I

Type: Primary
Antigen: SEMG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR