Anti-MAPT Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX54655
10794-456EA 615.99 USD
10794-456
Anti-MAPT Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Tau (phospho Thr217)

Type: Primary
Antigen: MAPT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR