Anti-STAU2 Mouse Monoclonal Antibody [clone: 1C6]

Supplier: GeneTex
GTX54653
10791-914EA 637.75 USD
10791-914
Anti-STAU2 Mouse Monoclonal Antibody [clone: 1C6]
Antibodies
Mouse Monoclonal antibody [1C6] to STAU2

Type: Primary
Antigen: STAU2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR