Anti-Cruciform DNA Mouse Monoclonal Antibody [clone: 2D3]

Supplier: GeneTex
GTX54648
10792-118EA 612.39 USD
10792-118
Anti-Cruciform DNA Mouse Monoclonal Antibody [clone: 2D3]
Antibodies
Mouse Monoclonal antibody [2D3] to Cruciform DNA

Type: Primary
Antigen: Cruciform DNA
Clonality: Monoclonal
Clone: 2D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR