Anti-PCK1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX54629
10792-898EA 610.88 USD
10792-898
Anti-PCK1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to PCK1

Type: Primary
Antigen: PCK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR