Anti-PAX2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX54602
10792-854EA 644.28 USD
10792-854
Anti-PAX2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to PAX2

Type: Primary
Antigen: PAX2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR