Anti-SDC1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX54537
10795-794EA 616.68 USD
10795-794
Anti-SDC1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Syndecan-1

Type: Primary
Antigen: SDC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR