Anti-Gabbr2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX54536
10795-792EA 496.3 USD
10795-792
Anti-Gabbr2 Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to GABA B Receptor 2

Type: Primary
Antigen: Gabbr2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR