Anti-HTT Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX54521
10794-204EA 616.68 USD
10794-204
Anti-HTT Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Huntingtin (neoepitope 552)

Type: Primary
Antigen: HTT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR