Anti-ICAM1 Mouse Monoclonal Antibody [clone: HM.1]

Supplier: GeneTex
GTX54507
10791-978EA 376.97 USD
10791-978
Anti-ICAM1 Mouse Monoclonal Antibody [clone: HM.1]
Mouse Monoclonal antibody [HM.1] to ICAM1

Type: Primary
Antigen: ICAM1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR