Anti-PTX3 Rat Monoclonal Antibody [clone: MNB4]

Supplier: GeneTex
GTX54484
10793-246EA 612.58 USD
10793-246
Anti-PTX3 Rat Monoclonal Antibody [clone: MNB4]
Antibodies
Rat Monoclonal antibody [MNB4] to Pentraxin 3

Type: Primary
Antigen: PTX3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR