Anti-IL6R Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX54364
10795-398EA 610.88 USD
10795-398
Anti-IL6R Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to IL6R

Type: Primary
Antigen: IL6R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR