Anti-egr2b Rabbit Polyclonal Antibody

GTX54231
10795-046EA 235.08 USD
10795-046
Anti-egr2b Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Egr2b

Type: Primary
Antigen: egr2b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Zebra Fish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR