Anti-bcl2a Rabbit Polyclonal Antibody

GTX54185
10795-996EA 235.08 USD
10795-996
Anti-bcl2a Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Bcl2

Type: Primary
Antigen: bcl2a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Zebra Fish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR