Anti-ctnnb2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX54176
10795-002EA 694.55 USD
10795-002
Anti-ctnnb2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Ctnnb1

Type: Primary
Antigen: ctnnb2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Zebra Fish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR