Anti-mmp14b Rabbit Polyclonal Antibody

GTX54170
10794-990EA 235.08 USD
10794-990
Anti-mmp14b Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Mmp14b

Type: Primary
Antigen: mmp14b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Zebra Fish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR