Anti-GALT Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX54159
10792-604EA 610.88 USD
10792-604
Anti-GALT Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to GALT

Type: Primary
Antigen: GALT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR