Anti-HTRA2 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX54154
10792-594EA 610.88 USD
10792-594
Anti-HTRA2 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to HtrA2

Type: Primary
Antigen: HTRA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR