Anti-CES1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX53943
10792-400EA 610.88 USD
10792-400
Anti-CES1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to CES1

Type: Primary
Antigen: CES1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR