Anti-tuba1c Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX128805
89424-634EA 588.94 USD
89424-634
Anti-tuba1c Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Tuba1l (tubulin, alpha 1, like)

Type: Primary
Antigen: tuba1c
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebra Fish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR