Anti-Nitroso-L Rat Polyclonal Antibody

GTX12878
10163-090EA 404.31 USD
10163-090
Anti-Nitroso-L Rat Polyclonal Antibody
Rat Polyclonal antibody to Nitroso-L.Cysteine

Type: Primary
Antigen: Nitroso-L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG
Reactivity: Other
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR