Anti-Influenza B Virus Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX128539
10166-804EA 577.14 USD
10166-804
Anti-Influenza B Virus Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Influenza B Virus NP

Type: Primary
Antigen: Influenza B Virus
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Influenza B virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR