Anti-mCherry Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX128508
89424-626EA 640.16 USD
89424-626
Anti-mCherry Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to mCherry

Type: Primary
Antigen: mCherry
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR